Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146258.1 | NP_001139730.1 | 3 | 466 | ||
NM_001146259.1 | NP_001139731.1 | 3 | 412 | |||
NM_001146260.1 | NP_001139732.1 | 3 | 466 | |||
NM_024779.4 | NP_079055.3 | 3 | 466 | |||
XM_005269152.2 | XP_005269209.1 | 3 | 459 | |||
XM_011538747.1 | XP_011537049.1 | 3 | 459 | |||
XM_017019973.1 | XP_016875462.1 | 3 | 423 | |||
GenBank mRNA | AK075553.1 | - | 3 | 422 | ||
AK125526.1 | - | 3 | 350 | |||
AK297243.1 | - | 3 | 451 | |||
AK302254.1 | - | 3 | 328 | |||
AK315588.1 | - | 3 | 346 | |||
AX754970.1 | - | 3 | 407 | |||
BC028596.2 | - | 3 | 351 | 4824916 | ||
BX419431.2 | - | 3 | 464 |