Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NHEJ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024782.2 | NP_079058.1 | 6 | 773 | ||
GenBank mRNA | AJ972687.1 | - | 6 | 717 | ||
AK022672.1 | - | 6 | 709 | |||
BC008210.2 | - | 2 | 99 | |||
BC012732.2 | - | 6 | 694 | 4026495 | ||
BC030986.1 | - | 6 | 726 | 4076851 | ||
CR457291.1 | - | 5 | 627 |