Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024786.2 | NP_079062.1 | 2 | 744 | ||
XM_017009869.1 | XP_016865358.1 | 3 | 460 | |||
XM_017009870.1 | XP_016865359.1 | 5 | 2271 | |||
XM_017009871.1 | XP_016865360.1 | 5 | 2220 | |||
XM_017009872.1 | XP_016865361.1 | 3 | 460 | |||
GenBank mRNA | AK023215.1 | - | 2 | 744 | ||
BC032000.1 | - | 2 | 363 | 4750082 |
Gene Symbol: ZDHHC11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010110.1 | XP_016865599.1 | 6 | 844 | ||
XM_017010111.1 | XP_016865600.1 | 5 | 671 | |||
XM_017010112.1 | XP_016865601.1 | 5 | 1248 | |||
XM_017010113.1 | XP_016865602.1 | 6 | 1370 | |||
XM_017010114.1 | XP_016865603.1 | 6 | 4131 | |||
XM_017010115.1 | XP_016865604.1 | 5 | 1196 | |||
XM_017010116.1 | XP_016865605.1 | 4 | 1596 | |||
XM_017010117.1 | XP_016865606.1 | 4 | 1596 | |||
XM_017010118.1 | XP_016865607.1 | 4 | 1596 | |||
XM_017010119.1 | XP_016865608.1 | 4 | 1596 | |||
XM_017010120.1 | XP_016865609.1 | 4 | 1596 | |||
XM_017010121.1 | XP_016865610.1 | 4 | 1596 | |||
XM_017010122.1 | XP_016865611.1 | 4 | 1596 |