Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CSPP1 SIRNA Assays ›
Gene Symbol:
CSPP1
Gene Name:
centrosome and spindle pole associated protein 1
Gene Aliases:
CSPP, JBTS21
Chromosome Location:
Chr.8: 67062417 - 67196614 on Build GRCh38
UniGene ID:
Hs.370147
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36482
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CSPP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001291339.1
NP_001278268.1
2
529
NM_024790.6
NP_079066.5
4
376
XM_005251305.3
XP_005251362.2
5
755
XM_006716474.2
XP_006716537.2
5
755
XM_006716477.2
XP_006716540.2
5
755
XM_011517598.1
XP_011515900.1
5
755
XM_011517599.1
XP_011515901.1
5
755
XM_011517600.1
XP_011515902.1
5
755
XM_011517601.1
XP_011515903.1
5
755
XM_011517602.1
XP_011515904.1
5
755
XM_011517603.1
XP_011515905.1
4
438
XM_011517607.1
XP_011515909.1
4
342
XM_011517608.1
XP_011515910.1
4
420
XM_011517609.1
XP_011515911.1
2
461
XM_017013847.1
XP_016869336.1
5
755
XM_017013848.1
XP_016869337.1
5
755
XM_017013849.1
XP_016869338.1
5
755
XM_017013850.1
XP_016869339.1
5
755
XM_017013851.1
XP_016869340.1
4
420
XM_017013852.1
XP_016869341.1
5
755
XM_017013853.1
XP_016869342.1
4
420
XM_017013854.1
XP_016869343.1
5
755
XM_017013855.1
XP_016869344.1
5
755
XM_017013856.1
XP_016869345.1
4
420
GenBank mRNA
AJ583433.1
-
2
529
AK096205.1
-
4
349
AK127108.1
-
4
316
AM156947.1
-
4
377
BX649147.1
-
4
377
DA245734.1
-
4
317
DB030075.1
-
2
463
Back To Top
More Information
Panther Classification:
Molecular Function -
centromere DNA-binding protein
Gene Ontology Categories:
Function(s)
positive regulation of cytokinesis
positive regulation of cell division
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0