Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TM4SF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024795.3 | NP_079071.2 | 4 | 504 | ||
XM_011511876.2 | XP_011510178.1 | 4 | 410 | |||
GenBank mRNA | AK026453.1 | - | 4 | 503 | ||
AY358671.1 | - | 4 | 496 | |||
BC035754.1 | - | 4 | 491 | 5760060 | ||
BC137256.1 | - | 4 | 490 | 9021253 | ||
BC137257.1 | - | 4 | 490 | 9021254 | ||
DA926038.1 | - | 4 | 504 |