Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004386.2 | NP_004377.2 | 4, 5 | 748 | ||
GenBank mRNA | AB210004.1 | - | 4, 5 | 692 | ||
AF026547.1 | - | 3, 4 | 650 |