Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024815.3 | NP_079091.3 | 2 | 294 | ||
XM_011544650.1 | XP_011542952.1 | 3 | 1025 | |||
GenBank mRNA | AK124446.1 | - | 3 | 1025 | ||
BC016902.1 | - | 2 | 306 | 3914234 | ||
DA015404.1 | - | 2 | 294 |