Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H14 SIRNA Assays ›
Gene Symbol:
ZC3H14
Gene Name:
zinc finger CCCH-type containing 14
Gene Aliases:
MSUT-2, NY-REN-37, SUT2, UKp68
Chromosome Location:
Chr.14: 88562909 - 88614720 on Build GRCh38
UniGene ID:
Hs.684046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36558
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H14
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001160103.1
NP_001153575.1
14
2162
NM_001160104.1
NP_001153576.1
14
2147
NM_001326295.1
NP_001313224.1
12
1769
NM_001326296.1
NP_001313225.1
13
2087
NM_001326297.1
NP_001313226.1
14
2343
NM_001326298.1
NP_001313227.1
11
1694
NM_001326299.1
NP_001313228.1
13
2072
NM_001326300.1
NP_001313229.1
13
2047
NM_001326301.1
NP_001313230.1
13
2268
NM_001326302.1
NP_001313231.1
13
2047
NM_001326303.1
NP_001313232.1
11
1875
NM_001326304.1
NP_001313233.1
11
1654
NM_001326305.1
NP_001313234.1
12
1972
NM_001326306.1
NP_001313235.1
10
1538
NM_001326307.1
NP_001313236.1
13
2087
NM_001326308.1
NP_001313237.1
11
1654
NM_001326309.1
NP_001313238.1
10
1579
NM_001326310.1
NP_001313239.1
14
2147
NM_001326311.1
NP_001313240.1
10
1579
NM_001326312.1
NP_001313241.1
13
2072
NM_001326313.1
NP_001313242.1
10
1538
NM_001326314.1
NP_001313243.1
12
1972
NM_001326315.1
NP_001313244.1
14
2343
NM_001326316.1
NP_001313245.1
14
2010
NM_024824.4
NP_079100.2
14
2162
NM_207660.3
NP_997543.1
11
1694
NM_207661.2
NP_997544.1
12
1617
NM_207662.3
NP_997545.2
4
991
NR_136936.1
13
2002
XM_005268067.4
XP_005268124.1
14
2142
XM_005268068.4
XP_005268125.1
13
2082
XM_005268069.4
XP_005268126.1
12
1764
XM_005268070.4
XP_005268127.1
12
1764
XM_005268071.4
XP_005268128.1
11
1674
XM_005268073.3
XP_005268130.1
6
1384
XM_006720257.2
XP_006720320.1
5
1309
XM_011537160.2
XP_011535462.1
14
2270
XM_011537161.2
XP_011535463.1
13
2001
XM_017021648.1
XP_016877137.1
13
2082
XM_017021649.1
XP_016877138.1
13
2067
XM_017021650.1
XP_016877139.1
13
2067
GenBank mRNA
AF155107.1
-
3
253
AF474376.1
-
3
857
AK021868.1
-
11
1541
AK090809.1
-
1
914
AK302136.1
-
11
1509
AK303123.1
-
13
1613
AK304275.1
-
14
2018
AL834215.1
-
12
1617
AY578060.1
-
14
2046
AY578061.1
-
11
1633
AY578062.1
-
12
1617
AY578063.1
-
6
695
BC011793.1
-
14
2028
4298961
BC023641.2
-
10
1668
BC027607.2
-
4
977
BX248265.1
-
10
1684
BX248281.1
-
6
763
BX248769.1
-
1
52
BX640716.1
-
14
2041
BX640876.1
-
3
355
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of mRNA stability
negative regulation of mRNA polyadenylation
protein binding
poly(A) binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline