Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCMBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256378.1 | NP_001243307.1 | 7, 8 | 1023 | ||
NM_001256379.1 | NP_001243308.1 | 7, 8 | 833 | |||
NM_024834.3 | NP_079110.1 | 7, 8 | 1023 | |||
XM_005270157.4 | XP_005270214.1 | 7, 8 | 1105 | |||
XM_011540170.1 | XP_011538472.1 | 7, 8 | 864 | |||
XM_017016663.1 | XP_016872152.1 | 7, 8 | 1105 | |||
XM_017016664.1 | XP_016872153.1 | 7, 8 | 864 | |||
GenBank mRNA | AK001928.1 | - | 3, 4 | 1195 | ||
AK023143.1 | - | 7, 8 | 894 | |||
AK094075.1 | - | 7, 8 | 827 | |||
BC004183.2 | - | 7, 8 | 989 | 2961492 | ||
BC007219.1 | - | 7, 8 | 989 | 2961492 | ||
CR457299.1 | - | 7, 8 | 724 | |||
KU178788.1 | - | 7, 8 | 724 | |||
KU178789.1 | - | 7, 8 | 693 |