Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 7 | 998 | ||
XM_005257689.2 | XP_005257746.1 | 7 | 974 | |||
XM_011525265.2 | XP_011523567.1 | 6 | 737 | |||
XM_011525266.1 | XP_011523568.1 | 6 | 860 | |||
XM_017025107.1 | XP_016880596.1 | 6 | 711 | |||
GenBank mRNA | AF126964.1 | - | 6 | 729 | ||
AF268387.1 | - | 7 | 918 | |||
AK024883.1 | - | 7 | 954 | |||
AK124053.1 | - | 1 | 1369 | |||
AK124084.1 | - | 1 | 914 | |||
AK290687.1 | - | 6 | 754 | |||
AK292161.1 | - | 7 | 967 | |||
AK301614.1 | - | 4 | 286 | |||
BC137491.1 | - | 7 | 864 | 9021491 |