Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300965.1 | NP_001287894.1 | 6 | 670 | ||
NM_001321.2 | NP_001312.1 | 6 | 695 | |||
GenBank mRNA | AB209321.1 | - | 3 | 4847 | ||
AY349166.1 | - | 6 | 631 | |||
BC000992.2 | - | 6 | 643 | 3448693 | ||
U46006.1 | - | 5 | 591 | |||
U57646.1 | - | 6 | 647 |