Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003119.3 | NP_003110.1 | 2 | 268 | ||
NM_199367.2 | NP_955399.1 | 2 | 268 | |||
XM_005256321.4 | XP_005256378.1 | 2 | 277 | |||
XM_006721264.3 | XP_006721327.1 | 2 | 277 | |||
XM_017023597.1 | XP_016879086.1 | 2 | 277 | |||
XM_017023598.1 | XP_016879087.1 | 2 | 277 | |||
XM_017023599.1 | XP_016879088.1 | 2 | 277 | |||
GenBank mRNA | BC007692.1 | - | 2 | 250 | ||
BC035929.2 | - | 1 | 51 | |||
BC036104.1 | - | 2 | 253 | 5274587 | ||
DA770435.1 | - | 2 | 525 | |||
HY006239.1 | - | 2 | 269 | |||
Y16610.1 | - | 2 | 250 |