Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199120.1 | NP_001186049.1 | 4 | 381 | ||
NM_001199121.1 | NP_001186050.1 | 4 | 357 | |||
NM_024839.2 | NP_079115.1 | 4 | 357 | |||
GenBank mRNA | AF212152.2 | - | 4 | 327 | ||
AJ504713.1 | - | 4 | 317 | |||
AJ504714.1 | - | 3 | 396 | |||
AJ504715.1 | - | 3 | 386 | |||
AJ504716.1 | - | 4 | 317 | |||
AK026291.1 | - | 4 | 326 | |||
BC011730.2 | - | 4 | 319 | 4422871 | ||
BG708140.1 | - | 4 | 385 | |||
BI598757.1 | - | 4 | 384 | |||
DA270919.1 | - | 4 | 357 |
Gene Symbol: TRIM39-RPP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199119.1 | NP_001186048.1 | 9 | 1368 | ||
GenBank mRNA | EU012025.1 | - | 9 | 1097 |