Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160167.1 | NP_001153639.1 | 3 | 623 | ||
NM_001160169.1 | NP_001153641.1 | 2 | 359 | |||
NM_024841.4 | NP_079117.3 | 4 | 609 | |||
GenBank mRNA | AK055042.1 | - | 3 | 510 | ||
BX640959.1 | - | 4 | 590 | |||
DA344861.1 | - | 3 | 510 | |||
DR001466.1 | - | 2 | 292 |