Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083600.2 | NP_001077069.1 | 3 | 329 | ||
NM_001083601.2 | NP_001077070.1 | 4 | 429 | |||
NM_001317093.1 | NP_001304022.1 | 3 | 303 | |||
NM_001317094.1 | NP_001304023.1 | 3 | 329 | |||
NM_001317095.1 | NP_001304024.1 | 2 | 213 | |||
NM_001317096.1 | NP_001304025.1 | 3 | 329 | |||
NM_001317097.1 | NP_001304026.1 | 3 | 329 | |||
NM_001317098.1 | NP_001304027.1 | 3 | 329 | |||
NM_024845.3 | NP_079121.1 | 3 | 329 | |||
GenBank mRNA | AK024216.1 | - | 3 | 236 | ||
AK092005.1 | - | 2 | 205 | |||
AK130792.1 | - | 3 | 236 | |||
AK225178.1 | - | 3 | 236 | |||
AK297219.1 | - | 3 | 232 | |||
AK298566.1 | - | 3 | 238 | |||
AK302361.1 | - | 3 | 194 | |||
AK311524.1 | - | 3 | 236 | |||
AL833594.1 | - | 4 | 306 | |||
AY358543.1 | - | 3 | 225 | |||
BC011267.1 | - | 2 | 142 | 4181472 | ||
BG822206.1 | - | 3 | 185 | |||
CR457324.1 | - | 2 | 126 | |||
DA736660.1 | - | 3 | 322 | |||
DA786716.1 | - | 3 | 302 | |||
DC339939.1 | - | 3 | 329 | |||
DC401343.1 | - | 4 | 429 |