Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160305.2 | NP_001153777.1 | 5 | 808 | ||
NM_024860.3 | NP_079136.2 | 6 | 736 | |||
NR_134583.1 | 6 | 736 | ||||
XM_011523336.1 | XP_011521638.1 | 5 | 799 | |||
GenBank mRNA | AK024801.1 | - | 6 | 692 | ||
AK127136.1 | - | 4 | 922 | |||
AK290495.1 | - | 5 | 770 | |||
AK309468.1 | - | 5 | 767 | |||
BC022451.1 | - | 5 | 770 | 4794299 | ||
BC110433.1 | - | 6 | 720 |