Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024867.3 | NP_079143.3 | 10 | 1606 | ||
NM_144722.3 | NP_653323.1 | 10 | 1606 | |||
XM_005248376.3 | XP_005248433.1 | 10 | 1711 | |||
XM_005248377.3 | XP_005248434.1 | 10 | 1711 | |||
XM_005248378.3 | XP_005248435.1 | 10 | 1700 | |||
XM_011514135.2 | XP_011512437.1 | 10 | 1716 | |||
XM_011514136.2 | XP_011512438.1 | 10 | 1715 | |||
XM_011514137.2 | XP_011512439.1 | 10 | 1715 | |||
XM_011514138.2 | XP_011512440.1 | 10 | 1716 | |||
XM_011514139.2 | XP_011512441.1 | 10 | 1715 | |||
XM_011514140.2 | XP_011512442.1 | 9 | 1450 | |||
XM_011514141.2 | XP_011512443.1 | 11 | 1914 | |||
XM_011514142.1 | XP_011512444.1 | 10 | 1711 | |||
XM_017009880.1 | XP_016865369.1 | 9 | 1547 | |||
XM_017009881.1 | XP_016865370.1 | 10 | 2570 | |||
XM_017009882.1 | XP_016865371.1 | 10 | 1704 | |||
GenBank mRNA | AK058124.1 | - | 10 | 1531 | ||
AL832607.1 | - | 3 | 514 | |||
EF553528.1 | - | 10 | 1556 |