Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128843.2 | NP_001122315.2 | 6 | 444 | ||
NM_001244764.1 | NP_001231693.1 | 6 | 484 | |||
NM_024873.5 | NP_079149.3 | 3 | 389 | |||
XM_011532256.2 | XP_011530558.1 | 7 | 717 | |||
XM_011532257.2 | XP_011530559.1 | 7 | 717 | |||
XM_017008619.1 | XP_016864108.1 | 7 | 717 | |||
XM_017008620.1 | XP_016864109.1 | 7 | 580 | |||
XM_017008621.1 | XP_016864110.1 | 6 | 627 | |||
XM_017008622.1 | XP_016864111.1 | 6 | 880 | |||
XM_017008623.1 | XP_016864112.1 | 6 | 490 | |||
XM_017008624.1 | XP_016864113.1 | 5 | 435 | |||
XM_017008625.1 | XP_016864114.1 | 4 | 968 | |||
GenBank mRNA | AF277289.1 | - | 3 | 378 | ||
AJ320534.1 | - | 3 | 378 | |||
AK290409.1 | - | 3 | 370 | |||
AK301058.1 | - | 6 | 444 | |||
AK303192.1 | - | 6 | 486 | |||
BC128408.1 | - | 3 | 377 | 40115867 | ||
BP335455.1 | - | 3 | 389 | |||
DC389224.1 | - | 6 | 444 | |||
EF585496.1 | - | 3 | 378 |