Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 13 | 1219 | ||
NM_024876.3 | NP_079152.3 | 14 | 1587 | |||
GenBank mRNA | AF052122.1 | - | 3 | 181 | ||
AK057199.1 | - | 4 | 1400 | |||
BC013114.1 | - | 14 | 1397 | 4342891 | ||
BC027473.1 | - | 13 | 1186 | 4997823 |