Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COQ8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142555.2 | NP_001136027.1 | 10, 11 | 962 | ||
NM_024876.3 | NP_079152.3 | 11, 12 | 1330 | |||
GenBank mRNA | AK057199.1 | - | 1, 2 | 1143 | ||
BC013114.1 | - | 11, 12 | 1140 | 4342891 | ||
BC027473.1 | - | 10, 11 | 929 | 4997823 |