Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024896.2 | NP_079172.2 | 12 | 2246 | ||
XM_005251587.1 | XP_005251644.1 | 10 | 1851 | |||
XM_017015139.1 | XP_016870628.1 | 13 | 2039 | |||
GenBank mRNA | AB058718.2 | - | 12 | 2123 | ||
AK093217.1 | - | 10 | 1422 | |||
AK127218.1 | - | 12 | 2235 | |||
AK172831.1 | - | 6 | 950 | |||
AL834191.1 | - | 11 | 1878 | |||
BC031630.1 | - | 4 | 610 | |||
BC136771.1 | - | 12 | 2220 | 9020761 | ||
BC136773.1 | - | 12 | 2220 | 9020763 | ||
BC144404.1 | - | 12 | 2220 |