Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271833.1 | NP_001258762.1 | 10 | 1237 | ||
NM_024901.4 | NP_079177.2 | 10 | 1305 | |||
XM_006710921.2 | XP_006710984.2 | 11 | 1259 | |||
XM_006710922.1 | XP_006710985.1 | 10 | 1463 | |||
XM_006710923.1 | XP_006710986.1 | 10 | 1451 | |||
XM_006710924.1 | XP_006710987.1 | 10 | 1228 | |||
XM_011542187.1 | XP_011540489.1 | 11 | 1270 | |||
XM_011542189.1 | XP_011540491.1 | 10 | 1478 | |||
XM_011542190.2 | XP_011540492.1 | 11 | 1138 | |||
XM_017002386.1 | XP_016857875.1 | 10 | 1014 | |||
XM_017002387.1 | XP_016857876.1 | 10 | 1030 | |||
XM_017002388.1 | XP_016857877.1 | 10 | 1264 | |||
GenBank mRNA | AK026110.1 | - | 10 | 1108 | ||
AK301583.1 | - | 9 | 1101 | |||
BC004557.2 | - | 10 | 1261 | 3957641 |