Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WLS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002292.3 | NP_001002292.3 | 2 | 597 | ||
NM_024911.6 | NP_079187.3 | 2 | 658 | |||
XM_011542191.2 | XP_011540493.1 | 2 | 581 | |||
XM_011542192.2 | XP_011540494.1 | 2 | 591 | |||
XM_017002390.1 | XP_016857879.1 | 2 | 425 | |||
GenBank mRNA | AB097018.1 | - | 2 | 580 | ||
AK074583.1 | - | 2 | 613 | |||
AK074984.1 | - | 2 | 580 | |||
AK309779.1 | - | 3 | 537 | |||
AY359035.1 | - | 2 | 640 | |||
BC007211.1 | - | 1 | 196 | |||
BC110826.1 | - | 2 | 559 | 6183768 | ||
BC137109.1 | - | 2 | 455 | 9021106 | ||
BC137113.1 | - | 2 | 455 | 9021110 | ||
BG701224.1 | - | 2 | 634 | |||
BX538320.1 | - | 2 | 597 | |||
DQ323735.1 | - | 2 | 627 |