Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030824.2 | NP_110451.1 | 5, 6 | 873 | ||
XM_006722908.3 | XP_006722971.1 | 4, 5 | 353 | |||
XM_017027316.1 | XP_016882805.1 | 3, 4 | 301 | |||
XM_017027317.1 | XP_016882806.1 | 3, 4 | 203 | |||
XM_017027318.1 | XP_016882807.1 | 3, 4 | 806 | |||
GenBank mRNA | AK024418.1 | - | 5, 6 | 845 | ||
AK295923.1 | - | 2, 3 | 166 |