Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 6 | 1045 | ||
NM_024913.4 | NP_079189.4 | 7 | 1261 | |||
XM_017012649.1 | XP_016868138.1 | 6 | 1737 | |||
GenBank mRNA | AK025639.1 | - | 4 | 406 | ||
AK055624.1 | - | 6 | 1385 | |||
AK225313.1 | - | 4 | 406 | |||
AK289978.1 | - | 7 | 1258 | |||
AL832619.2 | - | 7 | 1216 | |||
AL832638.2 | - | 7 | 1085 | |||
AY358220.1 | - | 6 | 837 | |||
BC017764.1 | - | 3 | 258 | |||
BC018685.1 | - | 6 | 796 | |||
BC030538.2 | - | 3 | 275 | 3919732 |