Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105533.1 | NP_001099003.1 | 15 | 2200 | ||
NM_024913.4 | NP_079189.4 | 16 | 2416 | |||
XM_017012649.1 | XP_016868138.1 | 15 | 2892 | |||
GenBank mRNA | AK025639.1 | - | 13 | 1561 | ||
AK225313.1 | - | 13 | 1561 | |||
AK289978.1 | - | 16 | 2413 | |||
AL832619.2 | - | 16 | 2371 | |||
AL832638.2 | - | 16 | 2240 | |||
AY358220.1 | - | 15 | 1992 | |||
BC018685.1 | - | 15 | 1951 | |||
BC030538.2 | - | 12 | 1430 | 3919732 |