Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145315.1 | NP_001138787.1 | 9, 10 | 1059 | ||
NM_001145316.1 | NP_001138788.1 | 9, 10 | 1251 | |||
NM_001145317.1 | NP_001138789.1 | 8, 9 | 738 | |||
NM_001145318.1 | NP_001138790.1 | 8, 9 | 1010 | |||
NM_024918.3 | NP_079194.3 | 9, 10 | 1164 | |||
XM_006723876.2 | XP_006723939.1 | 8, 9 | 733 | |||
XM_017028070.1 | XP_016883559.1 | 9, 10 | 1011 | |||
XM_017028071.1 | XP_016883560.1 | 9, 10 | 1233 | |||
XM_017028072.1 | XP_016883561.1 | 8, 9 | 921 | |||
GenBank mRNA | AK023408.1 | - | 10, 11 | 889 | ||
AK093031.1 | - | 9, 10 | 1159 | |||
AK290738.1 | - | 9, 10 | 1251 | |||
AK301671.1 | - | 8, 9 | 943 | |||
AK301840.1 | - | 8, 9 | 688 | |||
BC026011.1 | - | 8, 9 | 1264 | 4827570 | ||
BC058899.1 | - | 9, 10 | 1017 | 6063417 |