Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145315.1 | NP_001138787.1 | 4 | 555 | ||
NM_001145316.1 | NP_001138788.1 | 4 | 747 | |||
NM_001145317.1 | NP_001138789.1 | 3 | 234 | |||
NM_001145318.1 | NP_001138790.1 | 3 | 506 | |||
NM_024918.3 | NP_079194.3 | 4 | 660 | |||
XM_006723876.2 | XP_006723939.1 | 3 | 229 | |||
XM_017028070.1 | XP_016883559.1 | 4 | 543 | |||
XM_017028071.1 | XP_016883560.1 | 4 | 765 | |||
XM_017028072.1 | XP_016883561.1 | 3 | 453 | |||
GenBank mRNA | AK023408.1 | - | 5 | 386 | ||
AK093031.1 | - | 4 | 655 | |||
AK290738.1 | - | 4 | 747 | |||
AK301671.1 | - | 3 | 439 | |||
AK301840.1 | - | 3 | 184 | |||
BC026011.1 | - | 3 | 760 | 4827570 | ||
BC058899.1 | - | 4 | 513 | 6063417 |