Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122841.2 | NP_001116313.1 | 6, 7 | 721 | ||
NM_024919.4 | NP_079195.3 | 6, 7 | 871 | |||
NR_110312.1 | 9, 10 | 1425 | ||||
XM_011536137.1 | XP_011534439.1 | 7, 8 | 951 | |||
XM_011536138.1 | XP_011534440.1 | 7, 8 | 767 | |||
XM_011536140.1 | XP_011534442.1 | 6, 7 | 806 | |||
XM_011536141.1 | XP_011534443.1 | 6, 7 | 746 | |||
XM_011536142.2 | XP_011534444.1 | 6, 7 | 784 | |||
XM_011536143.1 | XP_011534445.1 | 6, 7 | 682 | |||
XM_011536145.1 | XP_011534447.1 | 6, 7 | 721 | |||
XM_011536146.2 | XP_011534448.1 | 2, 3 | 1574 | |||
XM_017011317.1 | XP_016866806.1 | 7, 8 | 951 | |||
XM_017011318.1 | XP_016866807.1 | 2, 3 | 1540 | |||
GenBank mRNA | AK074110.1 | - | 6, 7 | 2471 | ||
AK097179.1 | - | 6, 7 | 871 | |||
AK097579.1 | - | 6, 7 | 721 | |||
AK125963.1 | - | 9, 10 | 1397 | |||
AK303194.1 | - | 5, 6 | 525 | |||
AK303407.1 | - | 4, 5 | 579 | |||
AK316478.1 | - | 5, 6 | 525 | |||
AL133077.1 | - | 3, 4 | 424 | |||
BC137144.1 | - | 6, 7 | 829 | 9021141 |