Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031723.3 | NP_001026893.1 | 6 | 983 | ||
NM_001278310.1 | NP_001265239.1 | 5 | 811 | |||
XM_011532262.2 | XP_011530564.1 | 8 | 867 | |||
XM_011532263.2 | XP_011530565.1 | 6 | 1367 | |||
XM_011532265.2 | XP_011530567.1 | 5 | 1311 | |||
XM_017008628.1 | XP_016864117.1 | 8 | 1049 | |||
GenBank mRNA | AK023700.1 | - | 1 | 102 | ||
AK024343.1 | - | 4 | 510 | |||
AL833076.2 | - | 5 | 2855 | |||
AL833412.1 | - | 5 | 673 | |||
AY186740.1 | - | 8 | 862 | |||
AY358272.1 | - | 6 | 837 | |||
BC022248.1 | - | 6 | 852 | 4771526 |