Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SVEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153366.3 | NP_699197.3 | 35 | 6054 | ||
GenBank mRNA | AJ619977.1 | - | 35 | 5889 | ||
AY243503.1 | - | 35 | 5889 | |||
DV080389.1 | - | 4 | 478 |