Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024927.4 | NP_079203.3 | 13 | 2982 | ||
NR_073573.1 | 13 | 2957 | ||||
NR_073574.1 | 13 | 2329 | ||||
XM_017025113.1 | XP_016880602.1 | 14 | 2953 | |||
XM_017025114.1 | XP_016880603.1 | 14 | 2944 | |||
XM_017025115.1 | XP_016880604.1 | 14 | 3103 | |||
XM_017025116.1 | XP_016880605.1 | 13 | 2818 | |||
XM_017025117.1 | XP_016880606.1 | 14 | 2773 | |||
XM_017025118.1 | XP_016880607.1 | 13 | 2959 | |||
GenBank mRNA | AB208939.1 | - | 11 | 4265 | ||
AK024672.1 | - | 11 | 1750 | |||
AK096767.1 | - | 13 | 2329 | |||
AL137584.1 | - | 3 | 741 | |||
BC016062.1 | - | 13 | 2560 | 4909623 | ||
BC052978.1 | - | 13 | 2903 | 6473233 |