Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024940.7 | NP_079216.4 | 37 | 3932 | ||
XM_011544661.1 | XP_011542963.1 | 37 | 3890 | |||
XM_011544662.1 | XP_011542964.1 | 37 | 3890 | |||
XM_011544663.2 | XP_011542965.1 | 37 | 3890 | |||
XM_017013865.1 | XP_016869354.1 | 37 | 3742 | |||
XM_017013866.1 | XP_016869355.1 | 35 | 4847 | |||
XM_017013867.1 | XP_016869356.1 | 30 | 3280 | |||
XM_017013868.1 | XP_016869357.1 | 31 | 3203 | |||
GenBank mRNA | AK126249.1 | - | 12 | 1051 | ||
BC041761.2 | - | 6 | 513 | |||
BC137175.1 | - | 37 | 3790 | 9021172 | ||
BC137176.1 | - | 37 | 3790 | 9021173 | ||
CR749353.1 | - | 29 | 3073 | |||
CR933718.1 | - | 25 | 2506 |