Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOCK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024940.7 | NP_079216.4 | 44 | 4642 | ||
XM_011544661.1 | XP_011542963.1 | 44 | 4600 | |||
XM_011544662.1 | XP_011542964.1 | 44 | 4600 | |||
XM_011544663.2 | XP_011542965.1 | 44 | 4600 | |||
XM_017013865.1 | XP_016869354.1 | 44 | 4452 | |||
XM_017013866.1 | XP_016869355.1 | 42 | 5557 | |||
XM_017013867.1 | XP_016869356.1 | 37 | 3990 | |||
XM_017013868.1 | XP_016869357.1 | 38 | 3913 | |||
GenBank mRNA | AK126249.1 | - | 19 | 1761 | ||
BC041761.2 | - | 13 | 1223 | |||
BC137175.1 | - | 44 | 4500 | 9021172 | ||
BC137176.1 | - | 44 | 4500 | 9021173 | ||
BX648352.1 | - | 28 | 2734 | |||
CR933718.1 | - | 32 | 3216 |