Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024942.3 | NP_079218.2 | 5 | 1307 | ||
GenBank mRNA | AK023552.1 | - | 5 | 1163 | ||
BC032645.1 | - | 5 | 1147 | 5519534 |