Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM192A SIRNA Assays ›
Gene Symbol:
FAM192A
Gene Name:
family with sequence similarity 192 member A
Gene Aliases:
C16orf94, CDA018, CDA10, NIP30
Chromosome Location:
Chr.16: 57152466 - 57186242 on Build GRCh38
UniGene ID:
Hs.396740
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s36814
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM192A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024946.2
NP_079222.1
3
418
XM_005256156.4
XP_005256213.1
3
386
XM_005256158.3
XP_005256215.1
3
1473
XM_005256160.2
XP_005256217.1
3
390
XM_005256163.2
XP_005256220.1
4
527
XM_005256164.3
XP_005256221.1
4
472
XM_005256165.2
XP_005256222.1
3
447
XM_006721275.3
XP_006721338.2
3
1156
XM_011523343.2
XP_011521645.1
3
264
XM_017023681.1
XP_016879170.1
3
482
XM_017023682.1
XP_016879171.1
4
1591
XM_017023683.1
XP_016879172.1
4
1542
XM_017023684.1
XP_016879173.1
4
757
XM_017023685.1
XP_016879174.1
4
1506
XM_017023686.1
XP_016879175.1
5
1576
XM_017023687.1
XP_016879176.1
5
1551
XM_017023688.1
XP_016879177.1
5
1494
XM_017023689.1
XP_016879178.1
5
875
XM_017023690.1
XP_016879179.1
5
826
XM_017023691.1
XP_016879180.1
5
879
XM_017023692.1
XP_016879181.1
4
663
XM_017023693.1
XP_016879182.1
4
566
XM_017023694.1
XP_016879183.1
5
669
XM_017023695.1
XP_016879184.1
5
781
XM_017023696.1
XP_016879185.1
4
1164
XM_017023697.1
XP_016879186.1
4
549
XM_017023698.1
XP_016879187.1
4
1272
XM_017023699.1
XP_016879188.1
4
800
XM_017023700.1
XP_016879189.1
4
269
XM_017023701.1
XP_016879190.1
4
735
XM_017023702.1
XP_016879191.1
4
455
XM_017023703.1
XP_016879192.1
3
195
XM_017023704.1
XP_016879193.1
3
193
GenBank mRNA
AF212249.1
-
3
371
AF271785.1
-
3
371
AF356585.1
-
2
169
AK023291.1
-
3
318
AK025452.1
-
3
393
AL834421.1
-
3
230
BC020536.1
-
3
386
3849620
BC063409.1
-
3
326
BC071952.1
-
2
187
6305656
BI545617.1
-
3
423
DB101263.1
-
3
393
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline