Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308116.1 | NP_001295045.1 | 14 | 2778 | ||
NM_024947.3 | NP_079223.3 | 14 | 2763 | |||
XM_005247789.2 | XP_005247846.1 | 15 | 2841 | |||
XM_006713753.1 | XP_006713816.1 | 15 | 2810 | |||
XM_006713754.3 | XP_006713817.1 | 15 | 2932 | |||
XM_006713756.2 | XP_006713819.1 | 13 | 2583 | |||
XM_011513186.2 | XP_011511488.1 | 14 | 2742 | |||
XM_011513187.2 | XP_011511489.1 | 13 | 2694 | |||
XM_011513188.2 | XP_011511490.1 | 14 | 2682 | |||
XM_017007235.1 | XP_016862724.1 | 15 | 2829 | |||
XM_017007236.1 | XP_016862725.1 | 15 | 2817 | |||
XM_017007237.1 | XP_016862726.1 | 13 | 2634 | |||
XM_017007238.1 | XP_016862727.1 | 12 | 2535 | |||
XM_017007239.1 | XP_016862728.1 | 12 | 2382 | |||
GenBank mRNA | AF380154.1 | - | 14 | 2700 | ||
AF444193.1 | - | 13 | 2715 | |||
AJ320486.3 | - | 14 | 2757 | |||
AK304411.1 | - | 13 | 2579 | |||
BC131772.1 | - | 14 | 2742 | 40123356 | ||
EF560717.1 | - | 14 | 2738 |