Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308116.1 | NP_001295045.1 | 5 | 537 | ||
NM_024947.3 | NP_079223.3 | 5 | 522 | |||
XM_005247789.2 | XP_005247846.1 | 5 | 552 | |||
XM_005247791.3 | XP_005247848.1 | 5 | 552 | |||
XM_005247793.2 | XP_005247850.1 | 5 | 552 | |||
XM_006713753.1 | XP_006713816.1 | 5 | 521 | |||
XM_006713754.3 | XP_006713817.1 | 5 | 643 | |||
XM_006713757.3 | XP_006713820.1 | 5 | 552 | |||
XM_011513186.2 | XP_011511488.1 | 5 | 552 | |||
XM_011513187.2 | XP_011511489.1 | 5 | 552 | |||
XM_017007235.1 | XP_016862724.1 | 5 | 540 | |||
XM_017007236.1 | XP_016862725.1 | 5 | 528 | |||
XM_017007239.1 | XP_016862728.1 | 2 | 93 | |||
GenBank mRNA | AF380154.1 | - | 5 | 459 | ||
AF444193.1 | - | 4 | 474 | |||
AJ320486.3 | - | 5 | 516 | |||
AK023029.1 | - | 5 | 504 | |||
AK300151.1 | - | 5 | 503 | |||
BC131772.1 | - | 5 | 501 | 40123356 | ||
BX647868.1 | - | 5 | 497 | |||
EF560717.1 | - | 5 | 497 |