Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM188A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318330.1 | NP_001305259.1 | 10, 11 | 1131 | ||
NM_024948.3 | NP_079224.1 | 11, 12 | 1183 | |||
XM_005252600.2 | XP_005252657.1 | 10, 11 | 1077 | |||
XM_005252602.2 | XP_005252659.1 | 9, 10 | 1025 | |||
XM_006717508.2 | XP_006717571.1 | 10, 11 | 1079 | |||
XM_011519690.2 | XP_011517992.1 | 11, 12 | 1160 | |||
XM_011519691.2 | XP_011517993.1 | 11, 12 | 1160 | |||
XM_011519693.1 | XP_011517995.1 | 11, 12 | 1169 | |||
XM_011519694.1 | XP_011517996.1 | 12, 13 | 1224 | |||
XM_017016671.1 | XP_016872160.1 | 10, 11 | 1079 | |||
XM_017016672.1 | XP_016872161.1 | 13, 14 | 1316 | |||
XM_017016673.1 | XP_016872162.1 | 11, 12 | 1186 | |||
XM_017016674.1 | XP_016872163.1 | 9, 10 | 1027 | |||
XM_017016675.1 | XP_016872164.1 | 9, 10 | 996 | |||
XM_017016676.1 | XP_016872165.1 | 8, 9 | 944 | |||
GenBank mRNA | AB014761.1 | - | 11, 12 | 949 | ||
AF063600.1 | - | 6, 7 | 449 | |||
AF176916.1 | - | 10, 11 | 1093 | |||
AK023459.1 | - | 11, 12 | 1170 | |||
AL833873.1 | - | 11, 12 | 1145 | |||
BC020605.1 | - | 10, 11 | 1096 | |||
BC067799.1 | - | 11, 12 | 1169 | 5278533 | ||
CR457315.1 | - | 11, 12 | 949 |