Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAA25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024953.3 | NP_079229.2 | 9 | 861 | ||
XM_005253938.1 | XP_005253995.1 | 8 | 707 | |||
XM_006719606.2 | XP_006719669.1 | 9 | 1521 | |||
XM_017019977.1 | XP_016875466.1 | 7 | 584 | |||
GenBank mRNA | AB054990.1 | - | 9 | 803 | ||
AK023151.1 | - | 6 | 506 | |||
AK292485.1 | - | 9 | 834 | |||
AK307494.1 | - | 10 | 1010 | |||
AK309772.1 | - | 8 | 687 | |||
BC034357.1 | - | 9 | 815 | |||
BC065481.1 | - | 9 | 794 | |||
BC113585.1 | - | 9 | 861 | 8322637 | ||
BC113587.1 | - | 9 | 861 | 8322639 | ||
BX641140.1 | - | 9 | 816 | |||
BX647450.1 | - | 9 | 814 | |||
BX647992.1 | - | 6 | 462 |