Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PANK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324191.1 | NP_001311120.1 | 7 | 1946 | ||
NM_001324193.1 | NP_001311122.1 | 6 | 971 | |||
NM_024960.5 | NP_079236.3 | 6 | 1049 | |||
NM_153638.3 | NP_705902.2 | 6 | 1778 | |||
NM_153640.3 | NP_705904.1 | 6 | 1114 | |||
NR_136715.1 | 6 | 1635 | ||||
XM_005260835.3 | XP_005260892.1 | 6 | 1231 | |||
XM_005260836.4 | XP_005260893.3 | 6 | 1051 | |||
XM_011529364.2 | XP_011527666.1 | 5 | 1457 | |||
XM_017028077.1 | XP_016883566.1 | 6 | 1008 | |||
XM_017028078.1 | XP_016883567.1 | 6 | 2349 | |||
XM_017028079.1 | XP_016883568.1 | 6 | 919 | |||
GenBank mRNA | AF494409.1 | - | 6 | 1617 | ||
AK021791.1 | - | 6 | 1049 | |||
AK097796.1 | - | 6 | 1297 | |||
AL713654.1 | - | 4 | 397 | |||
BC009421.1 | - | 6 | 1151 | |||
BC065019.1 | - | 6 | 1060 | |||
BC093633.1 | - | 4 | 393 | |||
BC101569.1 | - | 4 | 392 | |||
BC101571.1 | - | 4 | 393 | |||
BC107724.1 | - | 6 | 958 |