Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRTM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134745.1 | NP_001128217.1 | 3 | 689 | ||
NM_001282924.1 | NP_001269853.1 | 3 | 619 | |||
NM_001282928.1 | NP_001269857.1 | 2 | 530 | |||
NM_024993.4 | NP_079269.4 | 3 | 689 | |||
XM_011533117.2 | XP_011531419.1 | 2 | 595 | |||
GenBank mRNA | AK126961.1 | - | 3 | 689 | ||
AY182030.1 | - | 1 | 274 | |||
AY358324.1 | - | 3 | 598 | |||
BC098267.1 | - | 2 | 372 | 40026833 | ||
BC098301.1 | - | 2 | 372 | 40026836 | ||
BC099738.1 | - | 3 | 523 | 40026837 | ||
BC110058.1 | - | 3 | 523 | 40026832 |