Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033505.1 | NP_001028677.1 | 2 | 227 | ||
NM_001033506.1 | NP_001028678.1 | 2 | 227 | |||
NM_001326.2 | NP_001317.1 | 2 | 227 | |||
GenBank mRNA | AK290836.1 | - | 2 | 227 | ||
AK297371.1 | - | 2 | 222 | |||
BC009792.1 | - | 2 | 209 | |||
BC059948.1 | - | 2 | 81 | |||
BC108319.1 | - | 2 | 201 | 6197136 | ||
BG435786.1 | - | 2 | 228 | |||
U15782.1 | - | 2 | 192 |