Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF606 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025027.3 | NP_079303.2 | 7 | 2624 | ||
XM_005259276.1 | XP_005259333.1 | 7 | 2484 | |||
GenBank mRNA | AB058755.1 | - | 7 | 2627 | ||
AF455357.1 | - | 8 | 3186 | |||
AK024322.1 | - | 1 | 846 | |||
AK293097.1 | - | 7 | 2094 | |||
AK294037.1 | - | 6 | 2008 | |||
BC037209.1 | - | 7 | 2098 | 4837545 |