Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP3K19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018044.2 | NP_001018054.1 | 7 | 3246 | ||
NM_001018046.2 | NP_001018056.1 | 8 | 1131 | |||
NM_001018047.2 | NP_001018057.1 | 8 | 987 | |||
NM_001282883.1 | NP_001269812.1 | 7 | 981 | |||
NM_001321177.1 | NP_001308106.1 | 10 | 3877 | |||
NM_025052.4 | NP_079328.3 | 9 | 3585 | |||
XM_011511891.2 | XP_011510193.1 | 9 | 3725 | |||
XM_011511892.1 | XP_011510194.1 | 12 | 4293 | |||
XM_011511894.2 | XP_011510196.1 | 9 | 3700 | |||
XM_011511896.2 | XP_011510198.1 | 9 | 3566 | |||
XM_011511897.2 | XP_011510199.1 | 9 | 3646 | |||
XM_017005003.1 | XP_016860492.1 | 12 | 4260 | |||
XM_017005004.1 | XP_016860493.1 | 11 | 4145 | |||
XM_017005005.1 | XP_016860494.1 | 10 | 3946 | |||
XM_017005006.1 | XP_016860495.1 | 10 | 3308 | |||
XM_017005007.1 | XP_016860496.1 | 9 | 3693 | |||
GenBank mRNA | AK026727.1 | - | 1 | 466 | ||
AK122935.1 | - | 10 | 3877 | |||
AK131216.1 | - | 3 | 1724 | |||
AK225579.1 | - | 1 | 466 | |||
AY574901.1 | - | 9 | 3585 | |||
AY574902.1 | - | 7 | 3246 | |||
AY574904.1 | - | 8 | 1131 | |||
AY574905.1 | - | 8 | 987 | |||
BC034417.1 | - | 2 | 632 | |||
BC137276.1 | - | 9 | 3584 | 9021273 | ||
BC137277.1 | - | 9 | 3584 | 9021274 | ||
BC144555.1 | - | 7 | 980 |