Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256599.1 | NP_001243528.1 | 3 | 634 | ||
NM_001256600.1 | NP_001243529.1 | 4 | 665 | |||
NM_001256601.1 | NP_001243530.1 | 3 | 465 | |||
NM_001282378.1 | NP_001269307.1 | 2 | 362 | |||
NM_001282379.1 | NP_001269308.1 | 3 | 624 | |||
NM_025058.4 | NP_079334.3 | 3 | 624 | |||
NR_046327.1 | 3 | 624 | ||||
NR_046329.1 | 4 | 683 | ||||
NR_104150.1 | 3 | 624 | ||||
XM_011510002.1 | XP_011508304.1 | 4 | 567 | |||
XM_017002393.1 | XP_016857882.1 | 4 | 738 | |||
GenBank mRNA | AK026882.1 | - | 3 | 535 | ||
AK096158.1 | - | 3 | 512 | |||
AK128139.1 | - | 4 | 683 | |||
AK225075.1 | - | 3 | 535 | |||
AK296300.1 | - | 4 | 661 | |||
AK297318.1 | - | 2 | 270 | |||
AK304542.1 | - | 3 | 468 | |||
AK308075.1 | - | 3 | 429 | |||
AK308637.1 | - | 2 | 664 | |||
AK309699.1 | - | 3 | 465 | |||
AK310429.1 | - | 4 | 770 | |||
AY251386.1 | - | 3 | 444 | |||
BC069416.1 | - | 3 | 520 | 7262300 | ||
BC069568.1 | - | 3 | 520 | 7262312 | ||
BC126372.1 | - | 3 | 541 | 8992088 | ||
BC143828.1 | - | 3 | 541 | |||
CN259033.1 | - | 2 | 314 | |||
DB262381.1 | - | 1 | 113 |