Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025059.3 | NP_079335.2 | 3 | 300 | ||
XM_011536147.2 | XP_011534449.1 | 3 | 338 | |||
XM_011536148.2 | XP_011534450.1 | 3 | 341 | |||
GenBank mRNA | AK026958.1 | - | 3 | 213 | ||
AK225330.1 | - | 3 | 214 | |||
BC060803.1 | - | 3 | 249 | 5298914 | ||
BI562667.1 | - | 3 | 254 | |||
DB232703.1 | - | 3 | 300 |