Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001268284.2 | NP_001255213.1 | 4 | 677 | ||
NM_001268285.2 | NP_001255214.1 | 2 | 391 | |||
NM_025061.5 | NP_079337.2 | 3 | 534 | |||
XM_011528319.2 | XP_011526621.1 | 3 | 1262 | |||
GenBank mRNA | AK091134.1 | - | 3 | 534 | ||
BC070089.1 | - | 3 | 534 | 30346745 | ||
BX538180.1 | - | 3 | 524 | |||
CD109368.1 | - | 3 | 561 | |||
CF593782.1 | - | 4 | 660 | |||
DA447942.1 | - | 2 | 356 |