Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UXS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253875.1 | NP_001240804.1 | 10 | 922 | ||
NM_001253876.1 | NP_001240805.1 | 5 | 1059 | |||
NM_025076.4 | NP_079352.2 | 10 | 907 | |||
NR_045607.1 | 9 | 858 | ||||
XM_006712775.3 | XP_006712838.1 | 10 | 884 | |||
XM_011511902.2 | XP_011510204.2 | 10 | 901 | |||
XM_011511903.2 | XP_011510205.2 | 10 | 884 | |||
XM_011511904.2 | XP_011510206.1 | 7 | 1229 | |||
XM_017005008.1 | XP_016860497.1 | 10 | 884 | |||
XM_017005009.1 | XP_016860498.1 | 10 | 869 | |||
XM_017005010.1 | XP_016860499.1 | 10 | 869 | |||
XM_017005011.1 | XP_016860500.1 | 10 | 1009 | |||
XM_017005012.1 | XP_016860501.1 | 10 | 944 | |||
XM_017005013.1 | XP_016860502.1 | 8 | 1467 | |||
XM_017005014.1 | XP_016860503.1 | 9 | 685 | |||
GenBank mRNA | AK027244.1 | - | 5 | 1059 | ||
AK075120.1 | - | 10 | 869 | |||
AK075170.1 | - | 10 | 884 | |||
AK122696.1 | - | 9 | 823 | |||
AK225592.1 | - | 5 | 1059 | |||
AK290668.1 | - | 10 | 882 | |||
AK304872.1 | - | 4 | 399 | |||
AY147934.1 | - | 10 | 874 | |||
AY358541.1 | - | 10 | 858 | |||
BC009819.2 | - | 10 | 848 | 3843312 |