Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025082.3 | NP_079358.3 | 14 | 1972 | ||
XM_006721277.2 | XP_006721340.1 | 12 | 1827 | |||
XM_011523344.1 | XP_011521646.1 | 13 | 1949 | |||
XM_011523345.1 | XP_011521647.1 | 5 | 950 | |||
XM_017023712.1 | XP_016879201.1 | 12 | 1826 | |||
XM_017023713.1 | XP_016879202.1 | 12 | 2254 | |||
XM_017023714.1 | XP_016879203.1 | 12 | 2359 | |||
XM_017023715.1 | XP_016879204.1 | 9 | 1699 | |||
XM_017023716.1 | XP_016879205.1 | 12 | 2620 | |||
XM_017023717.1 | XP_016879206.1 | 5 | 1062 | |||
GenBank mRNA | AK023173.1 | - | 10 | 1762 | ||
AK056097.1 | - | 14 | 1972 | |||
AK225399.1 | - | 11 | 1461 | |||
AK301621.1 | - | 11 | 1375 | |||
BC007642.1 | - | 11 | 1415 | |||
BC007864.2 | - | 2 | 221 | |||
BC015202.2 | - | 11 | 1423 | 3878214 | ||
BX648617.1 | - | 7 | 1402 | |||
CD242240.1 | - | 1 | 63 |